2-Hydroxypyridine ≥98.0%

Supplier: TCI America
Danger

Synonyms: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone

H0330-250G H0330-25G
TCH0330-250GEA 362 USD
TCH0330-250G TCH0330-025G
2-Hydroxypyridine ≥98.0%
2-Hydroxypyridine

CAS Number: 142-08-5 MDL Number: MFCD00006268 Molecular Formula: C5H5NO Molecular Weight: 95.10 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Boiling point (°C): 281 Melting point (°C): 107 Flash Point (°C): 161


Other Product Names / Synonyms: 2-Pyridinol, 2-Pyridone

Formula: C₅H₅NO
Boiling Pt: 281 °C
Melting Pt: 107 °C
Density: 1.085 g/cm³ (30 °C)
Flash Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD00006268
CAS Number: 142-08-5
EINECS: 205-520-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=121832894cc06a2488bae7682a1cbb964075997e&vid=121832894cc06a2488bae7682a1cbb964075997e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR