4-Hydroxy-4-methyl-2-pentanone ≥98.0% (by GC)

Supplier: TCI America

H0272-500ML
TCH0272-500MLEA 31.22 USD
TCH0272-500ML
4-Hydroxy-4-methyl-2-pentanone ≥98.0% (by GC)
4-Hydroxy-4-methyl-2-pentanone

CAS Number: 123-42-2 MDL Number: MFCD00004471 Molecular Formula: C6H12O2 Molecular Weight: 116.16 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 168 Melting point (°C): -44 Flash Point (°C): 58 Specific Gravity (20/20): 0.94 Lambda max.: 249 nm


Other Product Names / Synonyms: 4-Hydroxy-4-methyl-2-pentanone

Formula: (CH₃)₂C(OH)CH₂COCH₃
Boiling Pt: 168 °C
Melting Pt: –44 °C
Density: 0.94 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00004471
CAS Number: 123-42-2
UN: 1148
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4096f9fbd19d9dd51926c9e6d7aa323a14de8046&vid=4096f9fbd19d9dd51926c9e6d7aa323a14de8046&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR