Vanillin ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

H0264-25G H0264-500G
TCH0264-025GEA 41.89 USD
TCH0264-025G TCH0264-500G
Vanillin ≥98.0%
Vanillin

CAS Number: 121-33-5 MDL Number: MFCD00006942 Molecular Formula: C8H8O3 Molecular Weight: 152.15 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Boiling point (°C): 284 Melting point (°C): 83 Flash Point (°C): 158


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Others (Phytochemicals) Other Product Names / Synonyms: 4-Hydroxy-3-methoxybenzaldehyde

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 285
Melting Pt: 82
Density: 1.056 g/cm³ (20 °C)
Flash Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00006942
CAS Number: 121-33-5
EINECS: 204-465-2
Merck Index: 13,09998
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f20598fcce8d32293e2a36f74e9954a9b95543b&vid=03a1e9d52b7b6b3d94b449af19980b5f8635c304&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR