Hydroxyethyl cellulose, 200-300 mPa.s (2% in H₂0, 20 °C)

Supplier: TCI America

H0242-25G H0242-500G
TCH0242-25GEA 31.34 USD
TCH0242-25G TCH0242-500G
Hydroxyethyl cellulose, 200-300 mPa.s (2% in H₂0, 20 °C)
Hydroxyethyl cellulose

Form: Crystal
Color: Very Pale Yellow


Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Cellulose, Natural Polymers (for DDS)

MDL Number: MFCD00072770
CAS Number: 9004-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR