4-Hydroxyphenylarsonic acid 98,0

Supplier: TCI America

Synonyms: 4-Hydroxybenzenearsonic Acid

H0201-25G
TCH0201-025GEA 221.8 USD
TCH0201-025G
4-Hydroxyphenylarsonic acid 98,0
4-Hydroxyphenylarsonic acid

CAS Number: 98-14-6 MDL Number: MFCD00045707 Molecular Formula: C6H7AsO4 Molecular Weight: 218.04 Purity/Analysis Method: 98.0% (T) Form: Crystal Melting point (°C): 174


Other Product Names / Synonyms: 4-Hydroxybenzenearsonic Acid

Formula: C₆H₇AsO₄
MW: 218.04 g/mol
Melting Pt: 174 °C
MDL Number: MFCD00045707
CAS Number: 98-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=401fc162b4e3fbfebe21097a53ee477896202d30&vid=401fc162b4e3fbfebe21097a53ee477896202d30&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR