Hydroquinone ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Danger

H0186-500G H0186-25G
TCH0186-500GEA 43.9 USD
TCH0186-500G TCH0186-25G
Hydroquinone ≥99.0% (by titrimetric analysis)
Hydroquinone

CAS Number: 123-31-9 MDL Number: MFCD00002339 Molecular Formula: C6H6O2 Molecular Weight: 110.11 Purity/Analysis Method: 99.0% (T) Form: Crystal Boiling point (°C): 282 Melting point (°C): 173 Flash Point (°C): 165


Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Polymerization Inhibitors Other Product Names / Synonyms: 1,4-Dihydroxybenzene, Quinol, HQ

Formula: C₆H₄(OH)₂
Boiling Pt: 282 °C
Melting Pt: 173 °C
Density: 1.332 g/cm³ (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00002339
CAS Number: 123-31-9
EINECS: 204-617-8
UN: 3077
ADR: 9,III
Merck Index: 13,04833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1617bcf6caad784bc4042b9b3ab9740f32ad8544&vid=1617bcf6caad784bc4042b9b3ab9740f32ad8544&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR