Pentyl hexanoate ≥98.0%

Supplier: TCI America

H0106-25ML H0106-500ML
TCH0106-025MLEA 27 USD
TCH0106-025ML TCH0106-500ML
Pentyl hexanoate ≥98.0%

CAS Number: 540-07-8 MDL Number: MFCD00027281 Molecular Formula: C11H22O2 Molecular Weight: 186.30 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 226 Flash Point (°C): 91 Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Hexanoic Acid Amyl Ester, Pentyl Hexanoate, Hexanoic Acid Pentyl Ester

Formula: C₁₁H₂₂O₂
Boiling Pt: 226 °C
Melting Pt: –47 °C
Density: 0.87 (20 °C)
Flash Pt: 91 °C
MDL Number: MFCD00027281
CAS Number: 540-07-8
EINECS: 208-732-4
Merck Index: 13,00609
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc8b483373c2cb5e83369a65defd3325d860d7e6&vid=fc8b483373c2cb5e83369a65defd3325d860d7e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR