Cetrimonium chloride ≥95.0%

Supplier: TCI America
Warning

Synonyms: Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC

H0082-25G H0082-500G H0082-100G
TCH0082-025GEA 47.35 USD
TCH0082-025G TCH0082-500G TCH0082-100G
Cetrimonium chloride ≥95.0%
Cetrimonium chloride

CAS Number: 112-02-7 MDL Number: MFCD00011773 Molecular Formula: C19H42ClN Molecular Weight: 320.00 Purity/Analysis Method: 95.0% (T) Form: Crystal Melting point (°C): 234


Product Categories: Materials Chemistry, Cationic Surfactants, Bioscience, Quaternary Ammonium Salts for Clinical Chemistry Other Product Names / Synonyms: Cetyltrimethylammonium Chloride, HTAC, CTAC

Formula: C₁₉H₄₂ClN
Melting Pt: 234 °C
Density: 0.9 g/cm³ (20 °C)
MDL Number: MFCD00011773
CAS Number: 112-02-7
EINECS: 203-928-6
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0aa18485ed2f3e02b9ba4c0ad6701e4d2ce3b8d5&vid=0aa18485ed2f3e02b9ba4c0ad6701e4d2ce3b8d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR