Cetrimonium bromide ≥98.0% for synthesis
Supplier: TCI America
|
Danger
|
Synonyms:
N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
CAS Number: 57-09-0
MDL Number: MFCD00011772
Molecular Formula: C19H42BrN
Molecular Weight: 364.46
Purity/Analysis Method:
Product Categories: Materials Chemistry, Cationic Surfactants
Other Product Names / Synonyms: Cetyltrimethylammonium Bromide, CTAB
Formula:
CH₃(CH₂)₁₅N(Br)(CH₃)₃ Melting Pt: 243 °C Flash Pt: 244 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011772 CAS Number: 57-09-0 EINECS: 200-311-3 UN: 3077 ADR: 9,III Merck Index: 13,02034 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...