1-Hexadecanol ≥98.0%
Supplier: TCI America
CAS Number: 36653-82-4
MDL Number: MFCD00004760
Molecular Formula: C16H34O
Molecular Weight: 242.45
Purity/Analysis Method:
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Bioscience, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Cetanol, Cetyl Alcohol, Hexadecyl Alcohol
Formula:
H₃C(CH₂)₁₄CH₂OH Boiling Pt: 344 °C Melting Pt: 50 °C Density: 0.81 g/cm³ (20 °C) Flash Pt: 175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004760 CAS Number: 36653-82-4 EINECS: 253-149-0 Merck Index: 13,02037 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...