1-Heptene ≥98.0%

Supplier: TCI America
Danger

H0042-500ML H0042-25ML H0042-100ML
TCH0042-500MLEA 681.75 USD
TCH0042-500ML TCH0042-25ML TCH0042-100ML
1-Heptene ≥98.0%
1-Heptene

CAS Number: 592-76-7 MDL Number: MFCD00009531 Molecular Formula: C7H14 Molecular Weight: 98.19 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 94 Flash Point (°C): -4 Specific Gravity (20/20): 0.70

Formula: C₇H₁₄
Boiling Pt: 94 °C
Melting Pt: –119 °C
Density: 0.70 (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009531
CAS Number: 592-76-7
EINECS: 209-767-8
UN: 2278
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee3f441790be4c67fccad5a96a73b04477ebf962&vid=6591fd23f5cfbf248805009c2f77494a8582d604&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR