n-Heptane ≥99.0%

Supplier: TCI America
Danger

Synonyms: high purity solvents

H0027-100ML
TCH0027-100MLEA 48.18 USD
TCH0027-100ML
n-Heptane ≥99.0%
n-Heptane

[for Spectrophotometry]
CAS Number: 142-82-5
MDL Number: MFCD00009544
Molecular Formula: C7H16
Molecular Weight: 100.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Melting point (°C): -91
Flash Point (°C): -2
Specific Gravity (20/20): 0.68

Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98 °C
Melting Pt: –91 °C
Density: 0.68 (20 °C)
Flash Pt: –2 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2458cf2531f857d1972bdd582e0997b084a3558b&vid=f1789de25208039297558fbdc891548c95cb0771&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR