n-Hentriacontane ≥95.0% for synthesis

Supplier: TCI America

H0012-10G H0012-1G
TCH0012-010GEA 798 USD
TCH0012-010G TCH0012-001G
n-Hentriacontane ≥95.0% for synthesis
n-Hentriacontane

CAS Number: 630-04-6 MDL Number: MFCD00048745 Molecular Formula: C31H64 Molecular Weight: 436.85 Purity/Analysis Method: 95.0% (GC) Form: Crystal Boiling point (°C): 180 Melting point (°C): 66

Formula: H₃C(CH₂)₂₉CH₃
MW: 436.85 g/mol
Boiling Pt: 180 °C
Melting Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00048745
CAS Number: 630-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea918217945890b31b728c13c3c9ee1eeb13ee79&vid=a17f3477a237f3095536b5c74af55a094b076cff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR