Gnetol ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
(E)-2-(3,5-Dihydroxystyryl)benzene-1,3-diol, 2,3',5',6-Tetrahydroxy-trans-stilbene
G0371-100MG
TCG0371-100MGEA
213
USD
TCG0371-100MG
Gnetol ≥97.0% (by GC)
Gnetol
CAS Number: 86361-55-9
MDL Number: MFCD12405548
Molecular Formula: C14H12O4
Molecular Weight: 244.25
Purity/Analysis Method:
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 2,3',5',6-Tetrahydroxy-trans-stilbene
Formula:
C₁₄H₁₂O₄ MW: 244.25 g/mol Melting Pt: 271 °C |
MDL Number:
MFCD12405548 CAS Number: 86361-55-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...