Gemfibrozil ≥98.0%
Supplier: TCI America
Synonyms:
5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid, CI-719, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeric acid
CAS Number: 25812-30-0
MDL Number: MFCD00079335
Molecular Formula: C15H22O3
Molecular Weight: 250.34
Purity/Analysis Method:
Product Categories: Bioscience, Antilipemic Ingredients
Other Product Names / Synonyms: 2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic Acid, 2,2-Dimethyl-5-(2,5-dimethylphenoxy)valeric Acid
Formula:
C₁₅H₂₂O₃ MW: 250.34 Boiling Pt: 395 Melting Pt: 61 °C |
MDL Number:
MFCD00079335 CAS Number: 25812-30-0 Merck Index: 12,04394 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...