Glycine ≥99.0%

Supplier: TCI America

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

G0317-25G G0317-500G
TCG0317-25GEA 32 USD
TCG0317-25G TCG0317-500G
Glycine ≥99.0%
Glycine

CAS Number: 56-40-6 MDL Number: MFCD00008131 Molecular Formula: C2H5NO2 Molecular Weight: 75.07 Purity/Analysis Method: 99.0% (T) Form: Crystal Melting point (°C): 240


Product Categories: Analytical Science, Reagents for Gel & Buffer Preparation, etc. (Reagents for Electrophoresis) Other Product Names / Synonyms: Aminoacetic Acid, Glycocoll, H-Gly-OH

Formula: NH₂CH₂COOH
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 240 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a5b323cbbe2d463c584d51c654affa9af5ec666&vid=7353d07a229400bf7bf258bb7eeb5ea6cf641fe0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR