Glycerol-1,2-carbonate ≥90.0% (by GC)

Supplier: TCI America

G0279-100G
TCG0279-100GEA 359 USD
TCG0279-100G
Glycerol-1,2-carbonate ≥90.0% (by GC)
Glycerol-1,2-carbonate
Formula: C₄H₆O₄
Boiling Pt: 160°C
Flash Pt: 212°C
MDL Number: MFCD00085561
CAS Number: 931-40-8
EINECS: 213-235-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR