Genistein ≥98.0% (by HPLC)

Supplier: TCI America

G0272-1G G0272-100MG
TCG0272-001GEA 324.49 USD
TCG0272-001G TCG0272-100MG
Genistein ≥98.0% (by HPLC)
Genistein

CAS Number: 446-72-0 MDL Number: MFCD00016952 Molecular Formula: C15H10O5 Molecular Weight: 270.24 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Lambda max.: 262 nm (MeOH)Storage Temperature: 0-10°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals), Epigenetics Research Reagents Other Product Names / Synonyms: 4',5,7-Trihydroxyisoflavone

Formula: C₁₅H₁₀O₅
Melting Pt: 297…298 °C
MDL Number: MFCD00016952
CAS Number: 446-72-0
EINECS: 207-174-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f16d23a4205cc47fc4870e87676d4ee968722a61&vid=f16d23a4205cc47fc4870e87676d4ee968722a61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR