(E/Z)-Geranylacetone mixture E/Z ≥96.0%

Supplier: TCI America

Synonyms: 6,10-Dimethyl-5,9-undecadien-2-one, 6,10-Dimethyl-5,9-undecadien-2-one, mixture of isomers, Geranyl-2-propanone

G0236-25ML
TCG0236-025MLEA 50.88 USD
TCG0236-025ML
(E/Z)-Geranylacetone mixture E/Z ≥96.0%
(E/Z)-Geranylacetone

Geranylacetone [mixture of (E)- and (Z)- isomers, (3:2)]


Form: Clear Liquid
Specific Gravity (20/20): 0.87

Formula: (CH₃)₂C=CHCH₂CH₂C(CH₃)=CHCH₂CH₂C(O)CH₃
MW: 194.32 g/mol
Density: 0.87 (20 °C)
MDL Number: MFCD00008910
CAS Number: 689-67-8

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR