Tristearin (glycerol tristearate) ≥80.0%
Supplier: TCI America
Synonyms:
Propane-1,2,3-triyl tristearate, Tristearin
G0212-25G
G0212-250G
TCG0212-025GEA
99.96
USD
TCG0212-025G
TCG0212-250G
Tristearin (glycerol tristearate) ≥80.0%
Tristearin (Glycerol tristearate)
CAS Number: 555-43-1
MDL Number: MFCD00036230
Molecular Formula: C57H110O6
Molecular Weight: 891.50
Purity/Analysis Method:
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Tristearate
Formula:
C₅₇H₁₁₀O₆ Melting Pt: 73 °C |
MDL Number:
MFCD00036230 CAS Number: 555-43-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...