Guanidinium hydrochloride ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Warning

G0197-25G G0197-500G G0197-100G
TCG0197-025GEA 18.7 USD
TCG0197-025G TCG0197-500G TCG0197-100G
Guanidinium hydrochloride ≥99.0% (by titrimetric analysis)
Guanidinium hydrochloride

[for Protein Research] CAS Number: 50-01-1 MDL Number: MFCD00013026 Molecular Formula: CH5N3 Molecular Weight: 95.53 Purity/Analysis Method: 99.0% (T) Form: Crystal Melting point (°C): 187

Formula: NH₂C(=NH)NH₂·HCl
Melting Pt: 187 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52e4dd9f0a4b1455d358721a1fb2ac60050f650c&vid=2fe27eedee6de0a811023e1e664a52fd3375cc70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR