Guanine ≥98.0%

Supplier: TCI America

Synonyms: 2-Amino-1H-purin-6(7H)-one

G0169-25G G0169-250G G0169-1G
TCG0169-025GEA 35.73 USD
TCG0169-025G TCG0169-250G TCG0169-001G
Guanine ≥98.0%
Guanine

CAS Number: 73-40-5 MDL Number: MFCD00071533 Molecular Formula: C5H5N5O Molecular Weight: 151.13 Purity/Analysis Method: 98.0% (T,HPLC) Form: Crystal


Product Categories: Bioscience, Nucleobases and Their Analogs Other Product Names / Synonyms: 2-Amino-6-hydroxypurine, 2-Aminohypoxanthine

Formula: C₅H₅N₅O
Melting Pt: > 220 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071533
CAS Number: 73-40-5
EINECS: 200-799-8
Merck Index: 13,04580
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fae9141643408e2e2090c932a85d1a47f788636&vid=6fae9141643408e2e2090c932a85d1a47f788636&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR