2,5-Piperazinedione ≥98.0% (by total nitrogen basis)

Supplier: TCI America

G0100-500G G0100-25G
TCG0100-500GEA 747 USD
TCG0100-500G TCG0100-025G
2,5-Piperazinedione ≥98.0% (by total nitrogen basis)
2,5-Piperazinedione

CAS Number: 106-57-0 MDL Number: MFCD00006009 Molecular Formula: C4H6N2O2 Molecular Weight: 114.10 Purity/Analysis Method: 98.0% (N) Form: Crystal


Other Product Names / Synonyms: 2,5-Diketopiperazine, 2,5-Dioxopiperazine, Glycocoll Anhydride, 2,5-Piperazinedione

Formula: C₄H₆N₂O₂
MDL Number: MFCD00006009
CAS Number: 106-57-0
EINECS: 203-411-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4536299a0bd9595261ae6cb71d00a8de0185c0a0&vid=4536299a0bd9595261ae6cb71d00a8de0185c0a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR