Glycine ≥99.0%

Supplier: TCI America

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

G0099-500G G0099-25G
TCG0099-500GEA 46.82 USD
TCG0099-500G TCG0099-025G
Glycine ≥99.0%
Glycine

CAS Number: 56-40-6 MDL Number: MFCD00008131 Molecular Formula: C2H5NO2 Molecular Weight: 75.07 Purity/Analysis Method: 99.0% (T) Form: Crystal Melting point (°C): 240


Product Categories: Bioscience, Proteinogenic Amino Acids Other Product Names / Synonyms: Aminoacetic Acid, Glycocoll, H-Gly-OH

Formula: NH₂CH₂COOH
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 240 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a39b76541a75714e9d1ba7c24cc59d9eadbea407&vid=a39b76541a75714e9d1ba7c24cc59d9eadbea407&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR