Triolein ≥80.0%
Supplier: TCI America
CAS Number: 122-32-7
MDL Number: MFCD00137563
Molecular Formula: C57H104O6
Molecular Weight: 885.45
Purity/Analysis Method:
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol Trioleate
Formula:
C₅₇H₁₀₄O₆ Boiling Pt: 240 °C Melting Pt: –4 °C Density: 0.92 (20 °C) |
MDL Number:
MFCD00137563 CAS Number: 122-32-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...