ɑ,ɑ'-Dilaurin ≥96.0%
Supplier: TCI America
Synonyms:
Glycerol ɑ,ɑ'-Dilaurate
G0079-10G
TCG0079-010GEA
374.9
USD
TCG0079-010G
ɑ,ɑ'-Dilaurin ≥96.0%
alpha,alpha'-Dilaurin
CAS Number: 539-93-5
MDL Number: MFCD00048423
Molecular Formula: C27H52O5
Molecular Weight: 456.71
Purity/Analysis Method:
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol alpha,alpha'-Dilaurate
Formula:
C₂₇H₅₂O₅ MW: 456.71 g/mol Melting Pt: 59 °C |
MDL Number:
MFCD00048423 CAS Number: 539-93-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...