DL-Glutamine ≥98.0%
Supplier: TCI America
Synonyms:
H-DL-Gln-OH, 2,5-Diamino-5-oxopentanoic acid
G0062-10G
G0062-1G
TCG0062-010GEA
185.2
USD
TCG0062-010G
TCG0062-001G
DL-Glutamine ≥98.0%
DL-Glutamine
CAS Number: 6899-04-3
MDL Number: MFCD00065103
Molecular Formula: C5H10N2O3
Molecular Weight: 146.15
Purity/Analysis Method:
Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Gln-OH
Formula:
C₅H₁₀N₂O₃ |
MDL Number:
MFCD00065103 CAS Number: 6899-04-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...