L(+)-Glutamic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America

G0059-25G G0059-500G
TCG0059-025GEA 24.39 USD
TCG0059-025G TCG0059-500G
L(+)-Glutamic acid ≥99.0% (by titrimetric analysis)
L(+)-Glutamic acid

CAS Number: 56-86-0 MDL Number: MFCD00002634 Molecular Formula: C5H9NO4 Molecular Weight: 147.13 Purity/Analysis Method: 99.0% (T) Form: Crystal Melting point (°C): 225 Specific rotation [a]20/D: 32 deg (C=5, HCl(1+5))


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Bioscience, Proteinogenic Amino Acids Other Product Names / Synonyms: H-Glu-OH

Formula: HO₂CCH₂CH₂CH(NH₂)CO₂H
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 225 °C
Density: 1.538 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002634
CAS Number: 56-86-0
EINECS: 200-293-7
Merck Index: 13,04482
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=455d402740b3250b1bdaa1629d94ba3fffeb786d&vid=455d402740b3250b1bdaa1629d94ba3fffeb786d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR