β-D-Glucose ≥85.0% (contains alpha-D-Glucose)
Supplier: TCI America
CAS Number: 492-61-5
MDL Number: MFCD00063989
Molecular Formula: C6H12O6
Molecular Weight: 180.16
Purity/Analysis Method:
Formula:
C₆H₁₂O₆ Melting Pt: 155 °C |
MDL Number:
MFCD00063989 CAS Number: 492-61-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...