D-(+)-Glucosamine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 66-84-2
MDL Number: MFCD00135831
Molecular Formula: C6H13NO5
Molecular Weight: 215.63
Purity/Analysis Method:
Product Categories: Bioscience, Glucosamine, Aminosugars, Cosmetic Ingredients for Research and Experimental Use
Other Product Names / Synonyms: 2-Amino-2-deoxy-D-glucopyranose Hydrochloride
Formula:
C₆H₁₃NO₅ Melting Pt: 194 °C |
MDL Number:
MFCD00135831 CAS Number: 66-84-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...