Methyl-3,4,5-trihydroxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl gallate, Gallic acid methyl ester, Methyl 3,4,5-trihydroxybenzoate

G0017-500G G0017-25G G0017-100G
TCG0017-500GEA 222 USD
TCG0017-500G TCG0017-025G TCG0017-100G
Methyl-3,4,5-trihydroxybenzoate ≥98.0%
Methyl 3,4,5-trihydroxybenzoate

CAS Number: 99-24-1 MDL Number: MFCD00002194 Molecular Formula: C8H8O5 Molecular Weight: 184.15 Purity/Analysis Method: 98.0% (GC,T) Form: Crystal Melting point (°C): 200


Other Product Names / Synonyms: Gallic Acid Methyl Ester

Formula: C₈H₈O₅
MW: 184.15 g/mol
Boiling Pt: 450
Melting Pt: 200 °C
MDL Number: MFCD00002194
CAS Number: 99-24-1
EINECS: 202-741-7
Merck Index: 13,04366
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dcd980fd897e3c6309d5917ba0c366ac74b0f2a&vid=5dcd980fd897e3c6309d5917ba0c366ac74b0f2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR