Syringic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

G0014-500G G0014-25G G0014-100G
TCG0014-500GEA 521 USD
TCG0014-500G TCG0014-025G TCG0014-100G
Syringic acid ≥97.0% (by titrimetric analysis)
Syringic acid

CAS Number: 530-57-4 MDL Number: MFCD00002552 Molecular Formula: C9H10O5 Molecular Weight: 198.17 Purity/Analysis Method: 97.0% (T) Form: Crystal Melting point (°C): 209


Product Categories: Bioscience, Antioxidants, Others (Phytochemicals) Other Product Names / Synonyms: Gallic Acid 3,5-Dimethyl Ether, 4-Hydroxy-3,5-dimethoxybenzoic Acid

Formula: C₉H₁₀O₅
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 209 °C
Storage Temperature: Ambient
MDL Number: MFCD00002552
CAS Number: 530-57-4
EINECS: 208-486-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b314a349fad677268f3a1b80bd5f83eb07f8e6b5&vid=aa8d126ad0bfaa5dfc44a9b5e85b43c4e8089bd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR