Fullerene C70 ≥99.0% (by HPLC) for organic electronics

Supplier: TCI America

Synonyms: Buckminster-Fullerene C70, [5,6]-Fullerene-C70

F1233-100MG
TCF1233-100MGEA 369 USD
TCF1233-100MG
Fullerene C70 ≥99.0% (by HPLC) for organic electronics
Fullerene C70
Formula: C₇₀
MW: 840.77 g/mol
MDL Number: MFCD00282904
CAS Number: 115383-22-7

Order Now

Specification Test Results

Appearance Dark green to Dark blue to Black powder to crystal
Purity (HPLC) min. 99.0 area%
Electron Mobility (muFET) min. 0.30 cm2/Vs (OTS Si/SiO2 substrate)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR