2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)

Supplier: TCI America

F1217-1G F1217-5G
TCF1217-1GEA 55.62 USD
TCF1217-1G TCF1217-5G
2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)
2-Fluoro-4,4'-bis(trans-4-propylcyclohexyl)biphenyl
Formula: C₃₀H₄₁F
MDL Number: MFCD11053361
CAS Number: 102714-93-2
EINECS: 441-120-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a348cfae7f04c1d41a65b56298f54e024dd47dc4&vid=b8839439b8e1b520ae99e13fe9f8e26b1c68830d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR