5-Fluoro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

F1026-1G
TCF1026-1GEA 197 USD
TCF1026-1G
5-Fluoro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
5-Fluoro-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BFNO₂
MDL Number: MFCD07780754
CAS Number: 719268-92-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 55.0 to 59.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR