Ferrocenecarbaldehyde ≥97.0% (by GC, titration analysis)
Supplier: TCI America
F0998-25G
F0998-5G
TCF0998-25GEA
235
USD
TCF0998-25G
TCF0998-5G
Ferrocenecarbaldehyde ≥97.0% (by GC, titration analysis)
Ferrocenecarbaldehyde
Formula:
C₁₁H₁₀FeO |
MDL Number:
MFCD00001429 CAS Number: 12093-10-6 EINECS: 235-158-1 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Chelometric Titration) | min. 97.0 % |
Melting point | 114.0 to 121.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...