2-Fluoro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)

Supplier: TCI America

F0846-1G F0846-5G
TCF0846-1GEA 17 USD
TCF0846-1G TCF0846-5G
2-Fluoro-5-(trifluoromethyl)pyridine ≥98.0% (by GC)
2-Fluoro-5-(trifluoromethyl)pyridine

CAS Number: 69045-82-5 MDL Number: MFCD07437945 Molecular Formula: C6H3F4N Molecular Weight: 165.09 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 120 Flash Point (°C): 32 Specific Gravity (20/20): 1.40

Formula: C₆H₃F₄N
Boiling Pt: 120 °C
Density: 1.40 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD07437945
CAS Number: 69045-82-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69d91a685614b61d7934c9b360fa51ea625c5e16&vid=69d91a685614b61d7934c9b360fa51ea625c5e16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR