Famciclovir ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-[2-(2-Amino-9H-purin-9-yl)ethyl]-1,3-propanediol Diacetate, 2-(2-(2-Amino-9H-purin-9-yl)ethyl)propane-1,3-diyl diacetate
CAS Number: 104227-87-4
MDL Number: MFCD00866964
Molecular Formula: C14H19N5O4
Molecular Weight: 321.34
Purity/Analysis Method:
Product Categories: Bioscience, Antiviral Ingredients, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs, Prodrug Ingredients
Other Product Names / Synonyms: 2-[2-(2-Amino-9H-purin-9-yl)ethyl]-1,3-propanediol Diacetate
Formula:
C₁₄H₁₉N₅O₄ MW: 321.34 g/mol Melting Pt: 105 °C |
MDL Number:
MFCD00866964 CAS Number: 104227-87-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...