α,α,α,4-Tetrafluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 67515-55-3
MDL Number: MFCD00061294
Molecular Formula: C8H4F4O2
Molecular Weight: 208.11
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,4-Tetrafluoro-m-toluic Acid
Formula:
C₈H₄F₄O₂ Melting Pt: 115 °C |
MDL Number:
MFCD00061294 CAS Number: 67515-55-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...