N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 167015-11-4
MDL Number: MFCD00151922
Molecular Formula: C37H31NO4S
Molecular Weight: 585.72
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-S-trityl-D-cysteine, Fmoc-D-Cys(Trt)-OH
Formula:
C₃₇H₃₁NO₄S Melting Pt: 174 °C |
MDL Number:
MFCD00151922 CAS Number: 167015-11-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...