1-Formyl-4-piperidinecarboxylic acid ≥95.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 84163-42-8
MDL Number: MFCD03425206
Molecular Formula: C7H11NO3
Molecular Weight: 157.17
Purity/Analysis Method:
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-Formylisonipecotic Acid
Formula:
C₇H₁₁NO₃ Melting Pt: 138 °C |
MDL Number:
MFCD03425206 CAS Number: 84163-42-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...