4-Fluoro-2-nitrobenzoic acid ≥98.0% (by GC)
Supplier: TCI America
F0737-1G
F0737-5G
TCF0737-1GEA
53
USD
TCF0737-1G
TCF0737-5G
4-Fluoro-2-nitrobenzoic acid ≥98.0% (by GC)
4-Fluoro-2-nitrobenzoic acid
CAS Number: 394-01-4
MDL Number: MFCD00024300
Molecular Formula: C7H4FNO4
Molecular Weight: 185.11
Purity/Analysis Method:
Formula:
C₇H₄FNO₄ Melting Pt: 150 °C |
MDL Number:
MFCD00024300 CAS Number: 394-01-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...