Formylhydrazine ≥97.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: Formic acid hydrazide, Formohydrazide, Formic hydrazide

F0667-100G F0667-25G
TCF0667-100GEA 168 USD
TCF0667-100G TCF0667-25G
Formylhydrazine ≥97.0% (by HPLC)
Formylhydrazine
Formula: CH₄N₂O
MW: 60.06
Boiling Pt: 90
Melting Pt: 150
Storage Temperature: Refrigerator
MDL Number: MFCD00007589
CAS Number: 624-84-0
EINECS: 210-867-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 56.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR