2-Fluoroadenine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0647-1G F0647-200MG
TCF0647-1GEA 73 USD
TCF0647-1G TCF0647-200MG
2-Fluoroadenine ≥98.0% (by HPLC, titration analysis)
2-Fluoroadenine

CAS Number: 700-49-2
MDL Number: MFCD01632749
Molecular Formula: C5H4FN5
Molecular Weight: 153.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 6-Amino-2-fluoropurine

Formula: C₅H₄FN₅
MDL Number: MFCD01632749
CAS Number: 700-49-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf23a1ae4ca28592c35e2c9c11eab0d4877463d4&vid=20eeded4d0bb66b26cf7341e53bd75555edee823&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR