4-Fluorobenzyl mercaptan ≥95.0% (by GC, titration analysis)

Supplier: TCI America

F0612-5G
TCF0612-5GEA 97.3 USD
TCF0612-5G
4-Fluorobenzyl mercaptan ≥95.0% (by GC, titration analysis)
4-Fluorobenzyl mercaptan

CAS Number: 15894-04-9 MDL Number: MFCD00040724 Molecular Formula: C7H7FS Molecular Weight: 142.19 Purity/Analysis Method: 95.0% (GC,T) Form: Clear Liquid Boiling point (°C): 86 Flash Point (°C): 77 Specific Gravity (20/20): 1.17


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks Other Product Names / Synonyms: 4-Fluorobenzenemethanethiol, (4-Fluorophenyl)methanethiol

Formula: C₇H₇FS
Boiling Pt: 86 °C
Density: 1.17 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00040724
CAS Number: 15894-04-9
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=483e24017aeb36dbb4dc94e96b2a3ac63a0f3927&vid=483e24017aeb36dbb4dc94e96b2a3ac63a0f3927&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR