N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

F0596-25G F0596-5G
TCF0596-25GEA 176 USD
TCF0596-25G TCF0596-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate

CAS Number: 79990-15-1 MDL Number: MFCD00062960 Molecular Formula: C18H17NO4 Molecular Weight: 311.34 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 157 Specific rotation [a]20/D: 19 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected) Other Product Names / Synonyms: N-Fmoc-D-alanine, Fmoc-D-Ala-OH

Formula: C₁₈H₁₇NO₄
Melting Pt: 157 °C
MDL Number: MFCD00062960
CAS Number: 79990-15-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e63cd77833ce53114ee7ce06c4c63cd2b826529&vid=3e63cd77833ce53114ee7ce06c4c63cd2b826529&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR