N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 79990-15-1
MDL Number: MFCD00062960
Molecular Formula: C18H17NO4
Molecular Weight: 311.34
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-D-alanine, Fmoc-D-Ala-OH
Formula:
C₁₈H₁₇NO₄ Melting Pt: 157 °C |
MDL Number:
MFCD00062960 CAS Number: 79990-15-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...