α,α,α,4-Tetrafluoro-o-toluic acid ≥97.0%
Supplier: TCI America
F0577-5G
F0577-1G
TCF0577-5GEA
97
USD
TCF0577-5G
TCF0577-1G
α,α,α,4-Tetrafluoro-o-toluic acid ≥97.0%
α,α,α,4-Tetrafluoro-o-toluic acid
CAS Number: 141179-72-8
MDL Number: MFCD00040982
Molecular Formula: C8H4F4O2
Molecular Weight: 208.11
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha,alpha,alpha,4-Tetrafluoro-o-toluic Acid
Formula:
C₈H₄F₄O₂ MW: 208.11 Boiling Pt: 251 Melting Pt: 123 °C |
MDL Number:
MFCD00040982 CAS Number: 141179-72-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...