Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine ≥98.0%
Supplier: TCI America
Synonyms:
Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine
F0508-1G
F0508-5G
F0508-25G
TCF0508-1GEA
21
USD
TCF0508-1G
TCF0508-5G
TCF0508-25G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine ≥98.0%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine
CAS Number: 132388-59-1
Molecular Formula: C38H32N2O5
Molecular Weight: 596.68
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-Ngamma-trityl-L-asparagine, Fmoc-Asn(Trt)-OH
Formula:
C₃₈H₃₂N₂O₅ |
MDL Number:
MFCD00077049 CAS Number: 132388-59-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...