N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid ≥98.0%

Supplier: TCI America

Synonyms: N-Fmoc-L-aspartic Acid, Fmoc-Asp-OH

F0452-25G F0452-5G F0452-1G
TCF0452-25GEA 142 USD
TCF0452-25G TCF0452-5G TCF0452-1G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid ≥98.0%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid

CAS Number: 119062-05-4 MDL Number: MFCD00237654 Molecular Formula: C19H17NO6 Molecular Weight: 355.35 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Specific rotation [a]20/D: -30 deg (C=1, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected) Other Product Names / Synonyms: N-Fmoc-L-aspartic Acid, Fmoc-Asp-OH

Formula: C₁₉H₁₇NO₆
MW: 355.35 g/mol
MDL Number: MFCD00237654
CAS Number: 119062-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13fce049d5aa47d8da141fe86597f56a09d23a50&vid=13fce049d5aa47d8da141fe86597f56a09d23a50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR